Assay Details
| Gene Symbol: Tff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009362.2 | NP_033388.1 | 2 - 3 | 277 | 68 | |
| GenBank mRNA | AK008625.1 | - | 2 - 3 | 274 | 68 | |
| AK008809.1 | - | 2 - 3 | 274 | 68 | ||
| AK008873.1 | - | 2 - 3 | 277 | 68 | ||
| AK010179.1 | - | 2 - 3 | 274 | 68 | ||
| AK131896.1 | - | 2 - 3 | 278 | 68 | ||
| AK160174.1 | - | 2 - 3 | 274 | 68 | ||
| BC117054.1 | - | 2 - 3 | 274 | 40126373 | 68 | |
| BC117056.1 | - | 2 - 3 | 274 | 40126375 | 68 | |
| Z21858.1 | - | 2 - 3 | 246 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112005 | Z21858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112005 | NM_009362.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112005 | AK008809.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2112005 | AK131896.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021585 | Z21858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021585 | AK160174.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021585 | NM_009362.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021585 | AK008625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021585 | AK008809.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144185 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021585 | Z21858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021585 | AK160174.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021585 | NM_009362.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021585 | AK008625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021585 | AK008809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112005 | Z21858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112005 | NM_009362.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112005 | AK008809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2112005 | AK131896.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504884 | BC117056.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504884 | Z21858.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504884 | AK160174.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504884 | BC117054.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504884 | NM_009362.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138221 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144185 | - | - |
| Whole Mouse Genome G4122A | A_51_P504884 | BC117056.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504884 | Z21858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504884 | AK160174.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504884 | BC117054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P504884 | NM_009362.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114023 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13295224 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13295230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13295219 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13301194 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13295225 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13301182 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13295231 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13295221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13166091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13301180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13486459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13582327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13295221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13452027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138246 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13301185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13295224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13301194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13330419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13166083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13301184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138212 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13199288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P31138264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13183362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P13301182 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449667 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449668 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449669 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448156_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449666 | - | - |
| Mouse Genome 430 2.0 Array | 1448156_at | - | - |
| Mouse Genome 430 2.0 Array | 1448156_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700016666.mm.1 | NM_009362.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016666.mm.1 | NM_009362.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016666.mm.1 | TC1700001813.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700016667.mm.1 | NM_009362.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016667.mm.1 | NM_009362.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700016667.mm.1 | TC1700001813.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448156_at | - | - |
| Mouse Expression Set 430 | 1448156_PM_at | - | - |
| Murine Genome U74v2 Set | 160300_PM_at | - | - |
| Murine Genome U74v2 Set | 160300_at | - | - |
| Murine Genome U74v2 Set | 167147_at | - | - |
| Murine Genome U74v2 Set | 167147_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448156_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448156_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 457999 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3850452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2830821 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2830821 | - | - |
| mouse-6 v1.1 (Illumina) | GI_6678306-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | GI_6678306-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2830821 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2830821 | - | - |

Tff1
Genomic Map