| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800017580.mm.1 |
NM_027626.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017580.mm.1 |
NM_027626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017580.mm.1 |
NM_030263.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017580.mm.1 |
NM_030263.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017580.mm.1 |
NM_177698.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017580.mm.1 |
TC0800002339.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800017582.mm.1 |
NM_027626.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017582.mm.1 |
NM_027626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017582.mm.1 |
NM_030263.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017582.mm.1 |
NM_030263.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017582.mm.1 |
NM_177698.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017582.mm.1 |
TC0800002339.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800017579.mm.1 |
NM_027626.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017579.mm.1 |
NM_027626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017579.mm.1 |
NM_030263.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017579.mm.1 |
NM_030263.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017579.mm.1 |
NM_177698.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017579.mm.1 |
TC0800002339.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800017581.mm.1 |
NM_027626.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017581.mm.1 |
NM_027626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017581.mm.1 |
NM_030263.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017581.mm.1 |
NM_030263.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017581.mm.1 |
NM_177698.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800017581.mm.1 |
TC0800002339.mm.1 |
MTA TC |