Assay Details
| Gene Symbol: Psd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177698.4 | NP_808366.2 | 2 - 3 | 1511 | 86 | |
| GenBank mRNA | AB220685.1 | - | 2 - 3 | 1511 | 86 | |
| AK028684.1 | - | 3 - 4 | 1323 | 86 | ||
More Information
Set Membership: |
|

Psd3
Genomic Map