Assay Details
| Gene Symbol: Psd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030263.5 | NP_084539.2 | 3 - 4 | 425 | 118 | |
| NM_177698.4 | NP_808366.2 | 5 - 6 | 2186 | 118 | ||
| GenBank mRNA | AB220685.1 | - | 5 - 6 | 2186 | 118 | |
| AK028684.1 | - | 6 - 7 | 1998 | 118 | ||
| AK161498.1 | - | 3 - 4 | 411 | 118 | ||
| BC052365.1 | - | 3 - 4 | 389 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psd3
Genomic Map