Assay Details
| Gene Symbol: Psd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030263.5 | NP_084539.2 | 5 - 6 | 595 | 66 | |
| GenBank mRNA | AK028684.1 | - | 8 - 9 | 2168 | 66 | |
| AK161498.1 | - | 5 - 6 | 581 | 66 | ||
| BC052365.1 | - | 5 - 6 | 559 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psd3
Genomic Map