Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400022398.mm.1 |
TC0400002757.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022398.mm.1 |
NM_144904.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022398.mm.1 |
NM_144904.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022398.mm.1 |
NM_178164.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022398.mm.1 |
NM_178164.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022385.mm.1 |
TC0400002757.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022385.mm.1 |
NM_144904.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022385.mm.1 |
NM_144904.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022389.mm.1 |
TC0400002757.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400022389.mm.1 |
NM_144904.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022389.mm.1 |
NM_144904.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022389.mm.1 |
NM_178164.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400022389.mm.1 |
NM_178164.3 |
Refseq |