Assay Details
| Gene Symbol: Ptbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178164.3 | NP_835458.1 | 2 - 3 | 290 | 90 | |
| GenBank mRNA | AK033732.1 | - | 2 - 3 | 215 | 90 | |
| AK088560.1 | - | 2 - 3 | 222 | 90 | ||
| BC057641.1 | - | 2 - 3 | 195 | 4936557 | 90 | |
| CJ101924.1 | - | 2 - 3 | 291 | 90 | ||
| CJ103126.1 | - | 3 - 4 | 256 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033732.1, AK088560.1, BC057641.1, NM_178164.3 transcripts.
Set Membership: |
|

Ptbp3
Genomic Map