Assay Details
| Gene Symbol: Pigo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB038560.1 | - | 4 - 5 | 1621 | 61 | |
| AK050819.1 | - | 5 - 6 | 962 | 61 | ||
| AK220315.1 | - | 4 - 5 | 1569 | 61 | ||
| BC036977.1 | - | 5 - 6 | 966 | 61 | ||
| BC092394.1 | - | 4 - 5 | 1639 | 30534262 | 61 | |
| BC141058.1 | - | 4 - 5 | 1577 | 9055670 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigo
Genomic Map