Assay Details
| Gene Symbol: Pigo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB038560.1 | - | 7 - 8 | 3541 | 90 | |
| AK013913.1 | - | 1 - 2 | 92 | 90 | ||
| AK050819.1 | - | 8 - 9 | 2882 | 90 | ||
| AK220315.1 | - | 7 - 8 | 3489 | 90 | ||
| BC025857.1 | - | 2 - 3 | 1022 | 90 | ||
| BC036977.1 | - | 8 - 9 | 2814 | 90 | ||
| BC092394.1 | - | 7 - 8 | 3559 | 30534262 | 90 | |
| BC141058.1 | - | 7 - 8 | 3497 | 9055670 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigo
Genomic Map