Assay Details
Gene Symbol: Hint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026871.1 | NP_081147.1 | 4 - 5 | 402 | 65 | |
GenBank mRNA | AF356874.1 | - | 4 - 5 | 407 | 65 | |
AK004497.1 | - | 4 - 5 | 404 | 65 | ||
AK170135.1 | - | 3 - 4 | 1718 | 65 | ||
BC086940.1 | - | 4 - 5 | 402 | 6775318 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22872975 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22872977 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22872974 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22872979 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | BC086940.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | NM_026871.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | AF356874.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | AK004497.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | AK170135.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | BC086940.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | NM_026871.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | AF356874.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | AK004497.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P278887 | AK170135.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821510 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100619 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | BC086940.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | NM_026871.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | AF356874.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | AK004497.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | AK170135.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | BC086940.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | NM_026871.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | AF356874.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | AK004497.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P278887 | AK170135.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100619 | - | - |
Whole Mouse Genome G4122A | A_51_P278887 | BC086940.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | NM_026871.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | AF356874.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | AK004497.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | AK170135.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | BC086940.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | NM_026871.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | AF356874.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | AK004497.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P278887 | AK170135.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100619 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03547840 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03937862 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04088616 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04088614 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03547831 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04083221 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04088615 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22872977 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22872974 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22872975 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22872979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872944 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872947 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872963 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872957 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872937 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22873047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22872981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04088616 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03937861 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03294174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03294139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872974 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03547837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03294173 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03294145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04083221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04088612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03547831 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03547820 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03294177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22872971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04088614 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10512610 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512607 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512611 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512608 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10512609 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425196_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10512606 | - | - |
Mouse Genome 430 2.0 Array | 1425196_a_at | - | - |
Mouse Genome 430 2.0 Array | 1425196_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68917_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68917_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400020731.mm.1 | TC0400002536.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400020731.mm.1 | NM_026871.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400020731.mm.1 | NM_026871.1 | Refseq |
Mouse Expression Set 430 | 1425196_a_at | - | - |
Mouse Expression Set 430 | 1425196_PM_a_at | - | - |
Murine Genome U74v2 Set | 94365_at | - | - |
Murine Genome U74v2 Set | 94365_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425196_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1425196_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425196_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425196_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 451316 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5700086 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2759598 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2759598 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2759598 | - | - |
