Assay Details
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025900.2 | NP_080176.2 | 3 - 4 | 417 | 140 | |
GenBank mRNA | AK012615.1 | - | 3 - 4 | 358 | 140 | |
AK033451.1 | - | 3 - 4 | 307 | 140 | ||
AK154013.1 | - | 3 - 4 | 399 | 140 | ||
BC048844.1 | - | 3 - 4 | 367 | 140 | ||
BC055451.1 | - | 3 - 4 | 363 | 4164972 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
