Assay Details
Gene Symbol: Dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025900.2 | NP_080176.2 | 6 - 7 | 741 | 131 | |
GenBank mRNA | AK007546.1 | - | 3 - 4 | 290 | 131 | |
AK012615.1 | - | 6 - 7 | 682 | 131 | ||
AK012877.1 | - | 6 - 7 | 676 | 131 | ||
AK033451.1 | - | 6 - 7 | 631 | 131 | ||
BC055451.1 | - | 6 - 7 | 687 | 4164972 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
