Assay Details
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133985.2 | NP_598746.2 | 14 - 15 | 1671 | 102 | |
NR_045693.1 | 15 - 16 | 1800 | 102 | |||
GenBank mRNA | AK031790.1 | - | 13 - 14 | 1168 | 102 | |
AK075837.1 | - | 13 - 14 | 1179 | 102 | ||
AK129291.1 | - | 14 - 15 | 1666 | 102 | ||
BC036148.1 | - | 5 - 6 | 421 | 102 | ||
BC046453.1 | - | 15 - 16 | 1695 | 102 | ||
BC060645.1 | - | 14 - 15 | 1654 | 6852021 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
