Assay Details
| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008353.2 | NP_032379.2 | 11 - 12 | 1490 | 88 | |
| GenBank mRNA | AK137577.1 | - | 11 - 12 | 1491 | 88 | |
| BC057212.1 | - | 11 - 12 | 1473 | 5350447 | 88 | |
| U23922.1 | - | 11 - 12 | 1445 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il12rb1
Genomic Map