Assay Details
| Gene Symbol: Il12rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008353.2 | NP_032379.2 | 6 - 7 | 739 | 70 | |
| GenBank mRNA | AK137577.1 | - | 6 - 7 | 740 | 70 | |
| AK171937.1 | - | 6 - 7 | 716 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il12rb1
Genomic Map