Assay Details
| Gene Symbol: C78339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033192.2 | NP_001028364.1 | 3 - 4 | 984 | 97 | |
| GenBank mRNA | AK090371.1 | - | 3 - 4 | 368 | 97 | |
| AK141282.1 | - | 3 - 4 | 985 | 97 | ||
| AK171039.1 | - | 4 - 5 | 885 | 97 | ||
| AK218359.1 | - | 2 - 3 | 263 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C78339
Genomic Map