Assay Details
| Gene Symbol: C78339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033192.2 | NP_001028364.1 | 4 - 5 | 1117 | 120 | |
| GenBank mRNA | AK090371.1 | - | 4 - 5 | 501 | 120 | |
| AK141282.1 | - | 4 - 5 | 1118 | 120 | ||
| AK171039.1 | - | 5 - 6 | 1018 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009052 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009049 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009051 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009044 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009045 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009050 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009048 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982420 | AK141282.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982420 | AK171039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982420 | NM_001033192.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19638 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172140 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982420 | AK141282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982420 | AK171039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982420 | NM_001033192.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29009039 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172140 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07862973 | - | - |
| Whole Mouse Genome G4122A | A_51_P155550 | AK090371.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009044 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009051 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009050 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009049 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009045 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29009052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29009034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10610193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10683952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10868097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10949491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10954362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10954363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10745971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11119262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29009030 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404936 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433476_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433475_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404928 | - | - |
| Mouse Genome 430 2.0 Array | 1433475_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1433475_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1433476_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433476_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300003287.mm.1 | NM_001033192.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003287.mm.1 | NM_001033192.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003287.mm.1 | TC1300000536.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300003288.mm.1 | NM_001033192.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003288.mm.1 | NM_001033192.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300003288.mm.1 | TC1300000536.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433476_PM_at | - | - |
| Mouse Expression Set 430 | 1433475_a_at | - | - |
| Mouse Expression Set 430 | 1433475_PM_a_at | - | - |
| Mouse Expression Set 430 | 1433476_at | - | - |
| Murine Genome U74v2 Set | 97897_PM_at | - | - |
| Murine Genome U74v2 Set | 97897_at | - | - |
| Murine Genome U74v2 Set | 133906_at | - | - |
| Murine Genome U74v2 Set | 133906_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433475_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433476_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433475_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433476_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433476_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433475_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433475_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433476_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102030390 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3162465 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3162465 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85701695-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3162465 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3162465 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_85701695-S | - | - |

C78339
Genomic Map