Assay Details
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011390.2 | NP_035520.1 | 20 - 21 | 2805 | 99 | |
GenBank mRNA | AF087436.1 | - | 20 - 21 | 2807 | 99 | |
AK131129.1 | - | 20 - 21 | 2759 | 99 | ||
AK143535.1 | - | 20 - 21 | 2799 | 99 | ||
AK149750.1 | - | 20 - 21 | 2800 | 99 | ||
AK164405.1 | - | 3 - 4 | 1299 | 99 | ||
AK166215.1 | - | 20 - 21 | 2799 | 99 | ||
AK169950.1 | - | 20 - 21 | 2795 | 99 | ||
BC059242.1 | - | 20 - 21 | 2797 | 99 | ||
BC060133.1 | - | 20 - 21 | 2797 | 99 | ||
BC079677.1 | - | 16 - 17 | 4662 | 99 | ||
BC141107.1 | - | 20 - 21 | 2765 | 9055719 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
