Assay Details
| Gene Symbol: Tm6sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293795.1 | NP_001280724.1 | 6 - 7 | 679 | 134 | |
| GenBank mRNA | BC024498.1 | - | 6 - 7 | 667 | 4982620 | 134 |
More Information
Set Membership: |
|

Tm6sf2
Genomic Map