Assay Details
| Gene Symbol: Upf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122829.1 | NP_001116301.1 | 9 - 10 | 1561 | 66 | |
| NM_030680.2 | NP_109605.2 | 9 - 10 | 1528 | 66 | ||
| GenBank mRNA | AF182947.1 | - | 10 - 11 | 1517 | 66 | |
| AF322655.1 | - | 9 - 10 | 1248 | 66 | ||
| AK148196.1 | - | 10 - 11 | 1538 | 66 | ||
| BC052149.1 | - | 9 - 10 | 1466 | 6332288 | 66 | |
| BC056442.1 | - | 9 - 10 | 1561 | 5702493 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Upf1
Genomic Map