Assay Details
| Gene Symbol: Sugp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168290.1 | NP_001161762.1 | 4 - 5 | 2235 | 70 | |
| NM_172755.3 | NP_766343.3 | 4 - 5 | 2235 | 70 | ||
| GenBank mRNA | AF518875.1 | - | 4 - 5 | 2228 | 70 | |
| AK042293.1 | - | 4 - 5 | 2236 | 70 | ||
| AK122260.1 | - | 2 - 3 | 585 | 70 | ||
| AK137294.1 | - | 4 - 5 | 1839 | 70 | ||
| BC042763.1 | - | 2 - 3 | 583 | 70 | ||
| BC057305.1 | - | 4 - 5 | 1889 | 6822887 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sugp2
Genomic Map