Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900027735.mm.1 |
NM_173364.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027735.mm.1 |
NM_173364.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027735.mm.1 |
TC0900003293.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027742.mm.1 |
NM_173364.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027742.mm.1 |
NM_173364.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027742.mm.1 |
TC0900003293.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027738.mm.1 |
NM_173364.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027738.mm.1 |
NM_173364.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027738.mm.1 |
TC0900003293.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027736.mm.1 |
NM_173364.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027736.mm.1 |
NM_173364.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027736.mm.1 |
TC0900003293.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027739.mm.1 |
NM_173364.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027739.mm.1 |
NM_173364.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027739.mm.1 |
TC0900003293.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027741.mm.1 |
NM_173364.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027741.mm.1 |
NM_173364.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027741.mm.1 |
TC0900003293.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027737.mm.1 |
NM_173364.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027737.mm.1 |
NM_173364.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027737.mm.1 |
TC0900003293.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900027740.mm.1 |
NM_173364.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027740.mm.1 |
NM_173364.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900027740.mm.1 |
TC0900003293.mm.1 |
MTA TC |