| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300006343.mm.1 |
NM_175187.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006343.mm.1 |
NM_175187.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006343.mm.1 |
NR_029419.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006343.mm.1 |
NR_029419.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006343.mm.1 |
TC1300000983.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300006342.mm.1 |
NM_175187.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006342.mm.1 |
NM_175187.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006342.mm.1 |
NR_029418.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006342.mm.1 |
NR_029418.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006342.mm.1 |
NR_029419.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006342.mm.1 |
NR_029419.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300006342.mm.1 |
TC1300000983.mm.1 |
MTA TC |