Assay Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027704.2 | NP_081980.1 | 1 - 2 | 69 | 66 | |
| GenBank mRNA | AK016865.1 | - | 1 - 2 | 81 | 66 | |
| BC116803.1 | - | 1 - 2 | 69 | 40126122 | 66 | |
| BC116807.1 | - | 1 - 2 | 69 | 40126126 | 66 | |
| BC144831.1 | - | 1 - 2 | 69 | 66 | ||
| BC144832.1 | - | 1 - 2 | 69 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC116803.1, BC116807.1, NM_027704.2 transcripts.
Set Membership: |
|

Zdhhc11
Genomic Map