Assay Details
| Gene Symbol: Mtrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028628.1 | - | 9 - 10 | 1416 | 86 | |
| AK155359.1 | - | 10 - 11 | 86 | |||
| AK162978.1 | - | 8 - 9 | 1035 | 86 | ||
| AK163449.1 | - | 9 - 10 | 1398 | 86 | ||
| BC025942.1 | - | 5132457 | 86 | |||
| BC031565.1 | - | 9 - 10 | 86 | |||
| BC066796.1 | - | 8 - 9 | 1359 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtrr
Genomic Map