Assay Details
Gene Symbol: Prickle3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290624.1 | NP_001277553.1 | 1 - 2 | 120 | 121 | |
GenBank mRNA | AK037217.1 | - | 1 - 2 | 83 | 121 | |
BC023671.1 | - | 1 - 2 | 116 | 121 | ||
BC049258.1 | - | 1 - 2 | 62 | 5250488 | 121 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK037217.1, BC049258.1, NM_001290624.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
