Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00011138.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011138.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011138.mm.1 |
NM_016691.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011138.mm.1 |
NM_016691.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011138.mm.1 |
TC0X00001766.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011139.mm.1 |
NM_001243762.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011139.mm.1 |
NM_001243762.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011139.mm.1 |
NM_016691.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011139.mm.1 |
NM_016691.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011139.mm.1 |
TC0X00001766.mm.1 |
MTA TC |