Assay Details
| Gene Symbol: Glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053108.4 | NP_444338.2 | 2 - 3 | 417 | 96 | |
| GenBank mRNA | AB013137.1 | - | 2 - 3 | 383 | 96 | |
| AF109314.1 | - | 2 - 3 | 393 | 96 | ||
| AF276917.1 | - | 2 - 3 | 342 | 96 | ||
| AK013722.1 | - | 2 - 3 | 399 | 96 | ||
| AK153086.1 | - | 2 - 3 | 397 | 96 | ||
| BC012642.1 | - | 2 - 3 | 394 | 4206114 | 96 | |
| BC054418.1 | - | 2 - 3 | 379 | 96 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Glrx
Genomic Map