Assay Details
Gene Symbol: Ppm1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175523.4 | NP_780732.1 | 4 - 5 | 926 | 85 | |
GenBank mRNA | AK044610.1 | - | 4 - 5 | 996 | 85 | |
BC092238.1 | - | 4 - 5 | 926 | 30642218 | 85 | |
BC138473.1 | - | 3 - 4 | 732 | 8861493 | 85 | |
BC138474.1 | - | 3 - 4 | 732 | 8861494 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
