Assay Details
| Gene Symbol: Naf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163564.1 | NP_001157036.1 | 7 - 8 | 1479 | 81 | |
| GenBank mRNA | AK144735.1 | - | 7 - 8 | 726 | 81 | |
| BC027148.1 | - | 3 - 4 | 262 | 81 | ||
| BC053440.1 | - | 3 - 4 | 250 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2201822 | BC053440.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2201822 | NM_001163564.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2201822 | AK144735.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305529 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432939 | AK030532.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432939 | AK042380.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432939 | AK083365.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432939 | AK170119.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432939 | BC029235.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432939 | NM_197986.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432939 | NM_199145.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146873 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343351 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 659825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439791 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55971 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167334 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117962 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2201822 | BC053440.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2201822 | NM_001163564.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2201822 | AK144735.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167334 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06541168 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117962 | - | - |
| Whole Mouse Genome G4122A | A_51_P489086 | BC027148.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117962 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06772025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06970411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06772027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07095716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06895159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06987427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25934922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25935073 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572080 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452192_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572077 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395534 | - | - |
| Mouse Genome 430 2.0 Array | 1452192_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452192_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234344_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 234344_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800004631.mm.1 | NM_001163564.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004631.mm.1 | NM_001163564.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004631.mm.1 | TC0800000691.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452192_PM_at | - | - |
| Mouse Expression Set 430 | 1452192_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452192_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452192_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452192_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452192_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 662028 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2699013 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2699013 | - | - |

Naf1
Genomic Map