Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00018776.mm.1 |
NM_001289735.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018776.mm.1 |
NM_031259.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018776.mm.1 |
TC0X00003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018771.mm.1 |
NM_001289735.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018771.mm.1 |
NM_001289736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018771.mm.1 |
NM_031259.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018771.mm.1 |
TC0X00003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018774.mm.1 |
NM_001289736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018774.mm.1 |
TC0X00003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018772.mm.1 |
NM_001289735.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018772.mm.1 |
NM_001289736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018772.mm.1 |
NM_031259.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018772.mm.1 |
TC0X00003003.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018773.mm.1 |
NM_001289735.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018773.mm.1 |
NM_001289736.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018773.mm.1 |
NM_031259.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018773.mm.1 |
TC0X00003003.mm.1 |
MTA TC |