Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1432477_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432477_at |
- |
- |
Mouse Genome 430 2.0 Array |
1432477_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
73934_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
73934_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00018475.mm.1 |
NM_001281537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018475.mm.1 |
TC0X00002955.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018473.mm.1 |
NM_001281537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018473.mm.1 |
TC0X00002955.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018472.mm.1 |
NM_001281537.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018472.mm.1 |
TC0X00002955.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1432477_at |
- |
- |
Mouse Expression Set 430 |
1432477_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1432477_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1432477_at |
- |
- |