Assay Details
| Gene Symbol: Kif27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175214.3 | NP_780423.2 | 15 - 16 | 3551 | 146 | |
| GenBank mRNA | AK032912.1 | - | 8 - 9 | 1406 | 146 | |
| BC116646.1 | - | 13 - 14 | 2390 | 40086396 | 146 | |
More Information
Set Membership: |
|

Kif27
Genomic Map