Assay Details
| Gene Symbol: Dync1li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146229.2 | NP_666341.1 | 2 - 3 | 272 | 54 | |
| GenBank mRNA | AK082950.1 | - | 2 - 3 | 313 | 54 | |
| AK159702.1 | - | 314 | 54 | |||
| BC023347.1 | - | 2 - 3 | 272 | 5040044 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dync1li1
Genomic Map