Assay Details
| Gene Symbol: Crtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019922.2 | NP_064306.2 | 2 - 3 | 736 | 87 | |
| GenBank mRNA | AJ006469.1 | - | 2 - 3 | 694 | 87 | |
| AK007624.1 | - | 2 - 3 | 457 | 87 | ||
| AK017797.1 | - | 2 - 3 | 699 | 87 | ||
| AK047506.1 | - | 2 - 3 | 692 | 87 | ||
| BC049890.1 | - | 2 - 3 | 673 | 6306169 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crtap
Genomic Map