Assay Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB221628.1 | - | 3 - 4 | 1264 | 82 | |
AF191211.1 | - | 3 - 4 | 1264 | 82 | ||
BC068150.1 | - | 5 - 6 | 1851 | 82 | ||
BC128307.1 | - | 3 - 4 | 1259 | 82 | ||
BC128308.1 | - | 3 - 4 | 1260 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
