Assay Details
| Gene Symbol: Frrs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142965.1 | NP_001136437.1 | 4 - 5 | 833 | 80 | |
| GenBank mRNA | AK038461.1 | - | 4 - 5 | 537 | 80 | |
| AK078331.1 | - | 3 - 4 | 496 | 80 | ||
| BC132345.1 | - | 4 - 5 | 860 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092252 | AK078331.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092252 | BC132345.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092252 | NM_001142965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092252 | AK038461.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092251 | BC132345.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092251 | NM_001142965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2092251 | AK038461.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307559 | AK078331.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307559 | NM_001142965.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307559 | AK078331.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P307559 | NM_001142965.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179604 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665715 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91851 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191922 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867932 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187794 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165924 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152360 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104026 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307559 | AK078331.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307559 | NM_001142965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307559 | AK078331.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P307559 | NM_001142965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092251 | BC132345.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092251 | NM_001142965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092251 | AK038461.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092252 | AK078331.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092252 | BC132345.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092252 | NM_001142965.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2092252 | AK038461.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05247585 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152360 | - | - |
| Whole Mouse Genome G4122A | A_52_P440314 | AK078331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P440314 | BC132345.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P440314 | NM_001142965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P440314 | AK038461.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307559 | AK078331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307559 | NM_001142965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307559 | AK078331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307559 | NM_001142965.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128270 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03471652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03235541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03691932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03471647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04100880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22942705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03572208 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10513106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513111 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513108 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437640_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10513103 | - | - |
| Mouse Genome 430 2.0 Array | 1437640_at | - | - |
| Mouse Genome 430 2.0 Array | 1437640_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400021926.mm.1 | TC0400002712.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021926.mm.1 | NM_001142965.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021926.mm.1 | NM_001142965.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021925.mm.1 | TC0400002712.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400021925.mm.1 | NM_001142965.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400021925.mm.1 | NM_001142965.1 | Refseq |
| Mouse Expression Set 430 | 1437640_PM_at | - | - |
| Mouse Expression Set 430 | 1437640_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437640_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437640_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 475564 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216141 | - | - |

Frrs1l
Genomic Map