Assay Details
| Gene Symbol: Gm4035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079953.1 | - | 1 - 2 | 155 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lymphocyte antigen 86 | Ly86 |
MD-1, MD1 |
-11.0kb | Anti-sense | Chr. 13 - 37345345 - 37419036 on NCBI Build GRCm38 |
| Downstream | predicted gene 29590 | Gm29590 | 73.1kb | Anti-sense | Chr. 13 - 37508243 - 37512421 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm4035
Genomic Map