Assay Details
| Gene Symbol: Clcn4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035804.1 | - | 7 - 8 | 905 | 93 | |
| AK088317.1 | - | 5 - 6 | 666 | 93 | ||
| AK133759.1 | - | 7 - 8 | 931 | 93 | ||
| AK160325.1 | - | 6 - 7 | 1505 | 93 | ||
| BC005553.1 | - | 7 - 8 | 966 | 3586154 | 93 | |
| BC110668.1 | - | 8 - 9 | 1265 | 6391053 | 93 | |
| DQ186662.1 | - | 6 - 7 | 763 | 93 | ||
More Information
Set Membership: |
|

Clcn4-2
Genomic Map