Assay Details
| Gene Symbol: Clcn4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035804.1 | - | 8 - 9 | 1454 | 127 | |
| AK133759.1 | - | 9 - 10 | 1387 | 127 | ||
| BC005553.1 | - | 9 - 10 | 1422 | 3586154 | 127 | |
| BC110668.1 | - | 9 - 10 | 1814 | 6391053 | 127 | |
| DQ186662.1 | - | 7 - 8 | 1312 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clcn4-2
Genomic Map