Assay Details
| Gene Symbol: Nr2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010151.2 | NP_034281.2 | 1 - 2 | 865 | 71 | |
| GenBank mRNA | BC048695.1 | - | 1 - 2 | 551 | 71 | |
| BC108408.1 | - | 1 - 2 | 754 | 30245025 | 71 | |
| U07625.1 | - | 1 - 2 | 467 | 71 | ||
| X74134.1 | - | 1 - 2 | 765 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr2f1
Genomic Map