Assay Details
| Gene Symbol: Rsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081174.2 | NP_001074643.1 | 2 - 3 | 257 | 108 | |
| GenBank mRNA | BC147765.1 | - | 194 | 9087490 | 108 | |
| BC147767.1 | - | 194 | 9087492 | 108 | ||
More Information
Set Membership: |
|

Rsg1
Genomic Map