Assay Details
| Gene Symbol: Rsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081174.2 | NP_001074643.1 | 5 - 6 | 671 | 91 | |
| GenBank mRNA | BC147765.1 | - | 608 | 9087490 | 91 | |
| BC147767.1 | - | 608 | 9087492 | 91 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 60806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683568 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50453 | - | - |
| Whole Mouse Genome G4122A | A_52_P223654 | AK078152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P223654 | AK078152.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03711505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362768 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03386518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03378746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03711506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03729414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03925425 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03386514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03433712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03433711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362781 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10509945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509944 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430687_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10509941 | - | - |
| Mouse Genome 430 2.0 Array | 1430687_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430687_at | - | - |
| Mouse Expression Set 430 | 1430687_at | - | - |
| Mouse Expression Set 430 | 1430687_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430687_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430687_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215712 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3137189 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3137189 | - | - |

Rsg1
Genomic Map