Assay Details
Gene Symbol: Pax7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011039.2 | NP_035169.1 | 3 - 4 | 507 | 68 | |
GenBank mRNA | AF254422.4 | - | 3 - 4 | 507 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
