Assay Details
Gene Symbol: Htr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021358.2 | NP_067333.1 | 2 - 3 | 906 | 60 | |
GenBank mRNA | AF134158.1 | - | 2 - 3 | 906 | 60 | |
BC116652.1 | - | 2 - 3 | 40104888 | 60 | ||
BC116653.1 | - | 2 - 3 | 906 | 40104889 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
G-protein coupled receptor signaling pathway
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
serotonin receptor signaling pathway
learning
negative regulation of glutamate secretion
negative regulation of gamma-aminobutyric acid secretion
positive regulation of gamma-aminobutyric acid secretion
negative regulation of acetylcholine secretion, neurotransmission
cerebral cortex cell migration
positive regulation of TOR signaling
positive regulation of dopamine secretion
long-term synaptic potentiation
signal transducer activity
G-protein coupled receptor activity
G-protein coupled serotonin receptor activity
protein binding
signal transduction
G-protein coupled receptor signaling pathway
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
serotonin receptor signaling pathway
learning
negative regulation of glutamate secretion
negative regulation of gamma-aminobutyric acid secretion
positive regulation of gamma-aminobutyric acid secretion
negative regulation of acetylcholine secretion, neurotransmission
cerebral cortex cell migration
positive regulation of TOR signaling
positive regulation of dopamine secretion
long-term synaptic potentiation
signal transducer activity
G-protein coupled receptor activity
G-protein coupled serotonin receptor activity
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349530 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349529 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349525 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349536 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349524 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349533 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349528 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349522 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349538 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349535 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349526 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349537 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349531 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349534 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349523 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349532 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349527 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183497 | NM_021358.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183497 | AF134158.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183497 | NM_021358.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P183497 | AF134158.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503816 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157787 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 386701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392825 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610995 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237878 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135076 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172079 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139832 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183497 | NM_021358.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183497 | AF134158.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183497 | NM_021358.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P183497 | AF134158.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028350 | AK134977.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183497 | NM_021358.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183497 | AF134158.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183497 | NM_021358.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P183497 | AF134158.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349534 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349524 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349514 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349527 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349558 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349556 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349518 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349549 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349526 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349559 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349520 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349530 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349553 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349521 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349541 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349560 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349545 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349547 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349516 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349539 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349538 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349536 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349533 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349537 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349511 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349551 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349519 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349532 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349513 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23349542 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172079 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05424072 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139832 | - | - |
Whole Mouse Genome G4122A | A_51_P183497 | NM_021358.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183497 | AF134158.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183497 | NM_021358.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P183497 | AF134158.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110303 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172079 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139832 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03287456 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349523 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349535 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349534 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349531 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349527 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349538 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349525 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349528 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349537 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349530 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349533 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349524 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349522 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349526 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349536 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349532 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23349529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349501 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349485 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349553 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23349521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349539 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349508 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349551 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03959583 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03524463 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349560 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349541 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03818543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349549 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03484035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03326045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03426424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03641622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03955287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349516 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349563 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03287459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03326057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03919946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03433017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349502 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349565 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349452 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03287467 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349547 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03326051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349514 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03837543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349558 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03287449 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23349542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03426423 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10517675 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10517676 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10517674 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421757_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10517673 | - | - |
Mouse Genome 430 2.0 Array | 1421757_at | - | - |
Mouse Genome 430 2.0 Array | 1421757_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15565_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15565_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400031640.mm.1 | TC0400003839.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400031640.mm.1 | NM_021358.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400031640.mm.1 | NM_021358.2 | Refseq |
Mouse Expression Set 430 | 1421757_at | - | - |
Mouse Expression Set 430 | 1421757_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421757_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421757_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421757_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421757_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 543668 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4850022 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004850022 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219627 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1219627 | - | - |
mouse-6 v1.1 (Illumina) | scl23623.3.1_275-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4850022 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl23623.3.1_275-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850022 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2679840 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2679840 | - | - |
