Assay Details
| Gene Symbol: Taf7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028958.4 | NP_083234.2 | 2 - 3 | 363 | 131 | |
| GenBank mRNA | AF285574.1 | - | 2 - 3 | 167 | 131 | |
| AK017109.1 | - | 2 - 3 | 364 | 131 | ||
| BC106854.2 | - | 2 - 3 | 155 | 40046501 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf7l
Genomic Map