Assay Details
Gene Symbol: Was | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009515.2 | NP_033541.1 | 3 - 4 | 427 | 62 | |
GenBank mRNA | U29673.1 | - | 3 - 4 | 388 | 62 | |
U42471.1 | - | 3 - 4 | 388 | 62 | ||
U54788.1 | - | 3 - 4 | 388 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
