Assay Details
Gene Symbol: Epha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010139.3 | NP_034269.2 | 15 - 16 | 2803 | 125 | |
GenBank mRNA | AK137704.1 | - | 15 - 16 | 2785 | 125 | |
AK144202.1 | - | 15 - 16 | 2787 | 125 | ||
BC140960.1 | - | 15 - 16 | 2759 | 9055572 | 125 | |
X76010.1 | - | 8 - 9 | 1018 | 125 | ||
X78339.1 | - | 15 - 16 | 2765 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
