Assay Details
Gene Symbol: Epha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010139.3 | NP_034269.2 | 4 - 5 | 1115 | 98 | |
GenBank mRNA | AK137704.1 | - | 4 - 5 | 1097 | 98 | |
AK144202.1 | - | 4 - 5 | 1099 | 98 | ||
BC140960.1 | - | 4 - 5 | 1071 | 9055572 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
