Assay Details
| Gene Symbol: Epha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010139.3 | NP_034269.2 | 7 - 8 | 1717 | 64 | |
| GenBank mRNA | AK137704.1 | - | 7 - 8 | 1699 | 64 | |
| AK144202.1 | - | 7 - 8 | 1701 | 64 | ||
| BC140960.1 | - | 7 - 8 | 1673 | 9055572 | 64 | |
| X78339.1 | - | 7 - 8 | 1679 | 64 | ||
More Information
Set Membership: |
|

Epha2
Genomic Map